Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF38
Entrez Gene ID
152006
Gene Name
ring finger protein 38
Gene Aliases
-
Chromosome Location
Chr.9: 36336398 - 36487384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022781.4 NP_073618.3 4 - 5 764 85
NM_194328.2 NP_919309.1 4 - 5 931 85
NM_194329.2 NP_919310.1 3 - 4 614 85
NM_194330.2 NP_919311.1 4 - 5 662 85
NM_194332.2 NP_919313.1 3 - 4 781 85
XM_005251366.3 XP_005251423.1 4 - 5 768 85
XM_005251367.3 XP_005251424.1 4 - 5 666 85
XM_005251368.3 XP_005251425.1 4 - 5 780 85
XM_006716721.3 XP_006716784.1 3 - 4 628 85
XM_011517712.2 XP_011516014.1 5 - 6 704 85
XM_011517713.2 XP_011516015.1 4 - 5 768 85
XM_017014294.1 XP_016869783.1 3 - 4 685 85
XM_017014295.1 XP_016869784.1 4 - 5 795 85
XM_017014296.1 XP_016869785.1 3 - 4 365 85
XM_017014297.1 XP_016869786.1 3 - 4 361 85
GenBank mRNA AF394047.1 - 4 - 5 886 85
AK024996.1 - 2 - 3 274 85
AK093480.1 - 4 - 5 830 85
AK097653.1 - 4 - 5 824 85
AL136817.1 - 4 - 5 682 85
AV690059.2 - 4 - 5 451 85
AX748155.1 - 4 - 5 830 85
BC033786.1 - 3 - 4 614 85
DA483270.1 - 3 - 4 512 85

View Full Product Details