The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003995.3 | NP_003986.2 | 4 - 5 | 1120 | 74 |
XM_005251478.3 | XP_005251535.1 | 4 - 5 | 1907 | 74 | |
XM_005251479.4 | XP_005251536.1 | 6 - 7 | 5028 | 74 | |
XM_011517890.2 | XP_011516192.1 | 5 - 6 | 5612 | 74 | |
XM_011517891.2 | XP_011516193.1 | 6 - 7 | 5521 | 74 | |
XM_011517892.2 | XP_011516194.1 | 7 - 8 | 5693 | 74 | |
XM_011517893.2 | XP_011516195.1 | 6 - 7 | 5490 | 74 | |
XM_017014745.1 | XP_016870234.1 | 6 - 7 | 5024 | 74 | |
XM_017014746.1 | XP_016870235.1 | 6 - 7 | 5024 | 74 | |
XM_017014747.1 | XP_016870236.1 | 5 - 6 | 4478 | 74 | |
XM_017014748.1 | XP_016870237.1 | 6 - 7 | 5826 | 74 | |
XM_017014749.1 | XP_016870238.1 | 6 - 7 | 4986 | 74 | |
XM_017014750.1 | XP_016870239.1 | 5 - 6 | 4810 | 74 | |
GenBank mRNA | AJ005282.1 | - | 4 - 5 | 1120 | 74 |
L13436.1 | - | 4 - 5 | 1770 | 74 |