The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040633.1 | NP_001035723.1 | 5 - 6 | 1101 | 66 |
NM_001304531.1 | NP_001291460.1 | 2 - 3 | 123 | 66 | |
NM_016203.3 | NP_057287.2 | 5 - 6 | 1370 | 66 | |
NM_024429.1 | NP_077747.1 | 1 - 2 | 195 | 66 | |
XM_005250002.3 | XP_005250059.1 | 5 - 6 | 1236 | 66 | |
XM_005250004.3 | XP_005250061.1 | 5 - 6 | 1182 | 66 | |
XM_005250006.4 | XP_005250063.1 | 3 - 4 | 935 | 66 | |
XM_006716021.2 | XP_006716084.1 | 5 - 6 | 773 | 66 | |
XM_011516283.1 | XP_011514585.1 | 5 - 6 | 773 | 66 | |
XM_011516285.1 | XP_011514587.1 | 2 - 3 | 615 | 66 | |
XM_017012270.1 | XP_016867759.1 | 5 - 6 | 760 | 66 | |
XM_017012273.1 | XP_016867762.1 | 3 - 4 | 920 | 66 | |
GenBank mRNA | AB025580.1 | - | 1 - 2 | 108 | 66 |
AF087875.1 | - | 1 - 2 | 156 | 66 | |
AJ249976.1 | - | 5 - 6 | 849 | 66 | |
AK001887.1 | - | 1 - 2 | 195 | 66 | |
AK308116.1 | - | 5 - 6 | 801 | 66 | |
AK314646.1 | - | 1 - 2 | 157 | 66 | |
AW664063.1 | - | 2 - 3 | 95 | 66 | |
BC020540.1 | - | 1 - 2 | 76 | 66 | |
BC068598.1 | - | 5 - 6 | 1070 | 66 | |
BT007127.1 | - | 1 - 2 | 36 | 66 | |
DA151645.1 | - | 2 - 3 | 124 | 66 |