Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AFF1
Entrez Gene ID
4299
Gene Name
AF4/FMR2 family member 1
Gene Aliases
AF4, MLLT2, PBM1
Chromosome Location
Chr.4: 86935002 - 87141054 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166693.2 NP_001160165.1 16 - 17 3240 111
NM_001313959.1 NP_001300888.1 15 - 16 3364 111
NM_001313960.1 NP_001300889.1 13 - 14 2343 111
NM_005935.3 NP_005926.1 15 - 16 3351 111
XM_005263007.3 XP_005263064.1 16 - 17 3331 111
XM_005263009.3 XP_005263066.1 16 - 17 3232 111
XM_005263011.3 XP_005263068.1 15 - 16 3026 111
XM_005263013.3 XP_005263070.1 16 - 17 3235 111
XM_011531973.2 XP_011530275.1 16 - 17 3153 111
XM_017008215.1 XP_016863704.1 14 - 15 2197 111
XM_017008216.1 XP_016863705.1 12 - 13 2584 111
GenBank mRNA AK296243.1 - 13 - 14 2062 111
AK300364.1 - 16 - 17 3231 111
BC114930.1 - 15 - 16 3354 111
L13773.1 - 15 - 16 3364 111

View Full Product Details