Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AFF1
Entrez Gene ID
4299
Gene Name
AF4/FMR2 family member 1
Gene Aliases
AF4, MLLT2, PBM1
Chromosome Location
Chr.4: 86935002 - 87141054 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005935.3 NP_005926.1 18 - 19 3692 74
GenBank mRNA L13773.1 - 18 - 19 3705 74

View Full Product Details