The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001609.3 | NP_001600.1 | 5 - 6 | 797 | 84 |
XM_017016194.1 | XP_016871683.1 | 4 - 5 | 638 | 84 | |
GenBank mRNA | AK298638.1 | - | 4 - 5 | 626 | 84 |
AK314241.1 | - | 5 - 6 | 693 | 84 | |
AL831821.2 | - | 5 - 6 | 666 | 84 | |
BC013756.1 | - | 5 - 6 | 692 | 84 | |
KU177879.1 | - | 5 - 6 | 680 | 84 | |
KU177880.1 | - | 6 - 7 | 988 | 84 | |
U12778.1 | - | 5 - 6 | 695 | 84 |