The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022902.4 | NP_075053.2 | 10 - 11 | 1579 | 133 |
XM_005248569.3 | XP_005248626.1 | 9 - 10 | 1409 | 133 | |
XM_006714672.3 | XP_006714735.1 | 10 - 11 | 1573 | 133 | |
XM_017009749.1 | XP_016865238.1 | 9 - 10 | 1409 | 133 | |
GenBank mRNA | AF212235.1 | - | 2 - 3 | 258 | 133 |
AF461760.1 | - | 10 - 11 | 1473 | 133 | |
AK022558.1 | - | 7 - 8 | 943 | 133 | |
AK022818.1 | - | 8 - 9 | 1062 | 133 | |
AK292925.1 | - | 10 - 11 | 1443 | 133 | |
AL833716.1 | - | 2 - 3 | 333 | 133 | |
AY089991.1 | - | 10 - 11 | 1272 | 133 | |
AY358505.1 | - | 10 - 11 | 1490 | 133 | |
BC008198.1 | - | 7 - 8 | 963 | 133 | |
BX537394.1 | - | 10 - 11 | 1272 | 133 |