Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASB9P1
Entrez Gene ID
728619
Gene Name
ankyrin repeat and SOCS box containing 9 pseudogene 1
Gene Aliases
-
Chromosome Location
Chr.15: 92795484 - 92797108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033769.1 1 - 1 767 102

View Full Product Details