The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024334.2 | NP_077310.1 | 6 - 7 | 769 | 87 |
XM_017007176.1 | XP_016862665.1 | 7 - 8 | 891 | 87 | |
GenBank mRNA | AK027757.1 | - | 6 - 7 | 695 | 87 |
AK027827.1 | - | 1 - 2 | 87 | ||
AK027877.1 | - | 6 - 7 | 633 | 87 | |
AK074073.1 | - | 4 - 5 | 87 | ||
AK075010.1 | - | 6 - 7 | 620 | 87 | |
AK225589.1 | - | 6 - 7 | 87 | ||
AL136916.1 | - | 6 - 7 | 87 | ||
AY358625.1 | - | 6 - 7 | 532 | 87 | |
BC003125.1 | - | 6 - 7 | 664 | 87 | |
BC008054.2 | - | 4 - 5 | 259 | 87 | |
BC011719.2 | - | 6 - 7 | 87 | ||
KU178769.1 | - | 5 - 6 | 515 | 87 | |
KU178770.1 | - | 5 - 6 | 515 | 87 |