Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAX8
Entrez Gene ID
7849
Gene Name
paired box 8
Gene Aliases
-
Chromosome Location
Chr.2: 113215997 - 113278950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003466.3 NP_003457.1 5 - 6 641 92
NM_013952.3 NP_039246.1 5 - 6 641 92
NM_013953.3 NP_039247.1 5 - 6 641 92
NM_013992.3 NP_054698.1 5 - 6 641 92
XM_011511790.1 XP_011510092.1 5 - 6 673 92
XM_011511791.1 XP_011510093.1 5 - 6 673 92
XM_011511792.1 XP_011510094.1 5 - 6 673 92
XM_011511793.1 XP_011510095.1 5 - 6 673 92
XM_011511794.1 XP_011510096.1 5 - 6 642 92
XM_017004878.1 XP_016860367.1 5 - 6 673 92
XM_017004879.1 XP_016860368.1 5 - 6 642 92
GenBank mRNA AK225538.1 - 5 - 6 641 92
AK292191.1 - 5 - 6 638 92
AK309154.1 - 5 - 6 639 92
BC001060.2 - 5 - 6 632 92
KC733810.1 - 5 - 6 738 92
KU178404.1 - 4 - 5 475 92
KU178405.1 - 4 - 5 475 92
KU178406.1 - 3 - 4 277 92
L19606.1 - 4 - 5 485 92
S77904.1 - 5 - 6 635 92
S77905.1 - 5 - 6 635 92
S77906.2 - 5 - 6 635 92
X69699.1 - 5 - 6 635 92

View Full Product Details