Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL36A
Entrez Gene ID
27179
Gene Name
interleukin 36, alpha
Gene Aliases
FIL1, FIL1(EPSILON), FIL1E, IL-1F6, IL1(EPSILON), IL1F6
Chromosome Location
Chr.2: 113005461 - 113011071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014440.2 NP_055255.1 2 - 3 528 94
XM_005263639.2 XP_005263696.1 2 - 3 528 94
XM_011510965.1 XP_011509267.1 2 - 3 528 94
XM_017003806.1 XP_016859295.1 2 - 3 528 94
GenBank mRNA AF201831.1 - 1 - 2 117 94
AK314149.1 - 2 - 3 531 94
BC107042.1 - 1 - 2 117 94
BC107043.1 - 1 - 2 117 94

View Full Product Details