The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284401.1 | NP_001271330.1 | 6 - 7 | 1153 | 66 |
NM_001321294.1 | NP_001308223.1 | 7 - 8 | 1359 | 66 | |
NM_001321295.1 | NP_001308224.1 | 8 - 9 | 1508 | 66 | |
NR_135610.1 | 9 - 10 | 1630 | 66 | ||
XM_005264873.3 | XP_005264930.1 | 6 - 7 | 1153 | 66 | |
XM_017005725.1 | XP_016861214.1 | 6 - 7 | 1153 | 66 | |
XM_017005727.1 | XP_016861216.1 | 7 - 8 | 2293 | 66 | |
XM_017005728.1 | XP_016861217.1 | 7 - 8 | 1283 | 66 | |
GenBank mRNA | AK295839.1 | - | 6 - 7 | 1013 | 66 |
AL561480.3 | - | 2 - 3 | 66 | ||
BM678701.1 | - | 2 - 3 | 66 |