The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013415.1 | NP_001013433.1 | 4 - 5 | 689 | 76 |
NM_018315.4 | NP_060785.2 | 4 - 5 | 854 | 76 | |
NM_033632.3 | NP_361014.1 | 5 - 6 | 1028 | 76 | |
XM_011532083.1 | XP_011530385.1 | 7 - 8 | 2041 | 76 | |
XM_011532084.1 | XP_011530386.1 | 6 - 7 | 1991 | 76 | |
XM_011532085.1 | XP_011530387.1 | 5 - 6 | 1898 | 76 | |
XM_011532086.1 | XP_011530388.1 | 8 - 9 | 2219 | 76 | |
XM_011532087.1 | XP_011530389.1 | 7 - 8 | 2169 | 76 | |
XM_011532088.1 | XP_011530390.1 | 5 - 6 | 1427 | 76 | |
XM_017008362.1 | XP_016863851.1 | 7 - 8 | 2438 | 76 | |
GenBank mRNA | AB209725.1 | - | 4 - 5 | 618 | 76 |
AF383178.1 | - | 4 - 5 | 428 | 76 | |
AF411971.1 | - | 4 - 5 | 941 | 76 | |
AF411972.1 | - | 4 - 5 | 722 | 76 | |
AK001933.1 | - | 4 - 5 | 479 | 76 | |
AK223449.1 | - | 4 - 5 | 656 | 76 | |
AK294568.1 | - | 3 - 4 | 415 | 76 | |
AK315142.1 | - | 4 - 5 | 636 | 76 | |
AY008274.1 | - | 4 - 5 | 691 | 76 | |
AY033553.1 | - | 4 - 5 | 722 | 76 | |
AY049984.1 | - | 4 - 5 | 935 | 76 | |
BC037320.2 | - | 4 - 5 | 609 | 76 | |
BC117244.1 | - | 5 - 6 | 996 | 76 | |
BC117246.1 | - | 5 - 6 | 996 | 76 | |
BC143944.1 | - | 6 - 7 | 1046 | 76 | |
CR749305.1 | - | 5 - 6 | 1159 | 76 |