The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252119.1 | NP_001239048.1 | 15 - 16 | 3688 | 70 |
NM_001252120.1 | NP_001239049.1 | 16 - 17 | 3820 | 70 | |
NM_001252122.1 | NP_001239051.1 | 15 - 16 | 3562 | 70 | |
NM_015148.3 | NP_055963.2 | 15 - 16 | 3667 | 70 | |
XM_005246991.1 | XP_005247048.1 | 12 - 13 | 3002 | 70 | |
XM_011510826.1 | XP_011509128.1 | 15 - 16 | 3643 | 70 | |
XM_011510828.1 | XP_011509130.1 | 16 - 17 | 3744 | 70 | |
XM_011510829.1 | XP_011509131.1 | 15 - 16 | 3685 | 70 | |
XM_011510830.1 | XP_011509132.1 | 15 - 16 | 3686 | 70 | |
XM_011510831.2 | XP_011509133.1 | 13 - 14 | 3850 | 70 | |
XM_011510832.2 | XP_011509134.1 | 13 - 14 | 3829 | 70 | |
XM_011510833.1 | XP_011509135.1 | 12 - 13 | 3023 | 70 | |
XM_011510834.1 | XP_011509136.1 | 8 - 9 | 2746 | 70 | |
XM_011510835.1 | XP_011509137.1 | 8 - 9 | 2336 | 70 | |
XM_017003634.1 | XP_016859123.1 | 15 - 16 | 3911 | 70 | |
XM_017003635.1 | XP_016859124.1 | 16 - 17 | 3723 | 70 | |
XM_017003636.1 | XP_016859125.1 | 15 - 16 | 3679 | 70 | |
GenBank mRNA | AF387103.1 | - | 15 - 16 | 3607 | 70 |
AK296945.1 | - | 12 - 13 | 3172 | 70 | |
AK302417.1 | - | 15 - 16 | 3518 | 70 | |
AK302561.1 | - | 12 - 13 | 3002 | 70 | |
BC020054.1 | - | 11 - 12 | 2842 | 70 | |
BC043495.1 | - | 9 - 10 | 2424 | 70 | |
BC063585.1 | - | 15 - 16 | 3610 | 70 | |
CR749231.1 | - | 16 - 17 | 3821 | 70 | |
D50925.2 | - | 15 - 16 | 3619 | 70 | |
U79240.1 | - | 6 - 7 | 941 | 70 |