Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMARCD2
Entrez Gene ID
6603
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2
Gene Aliases
BAF60B, CRACD2, PRO2451, Rsc6p
Chromosome Location
Chr.17: 63832081 - 63842991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098426.1 NP_001091896.1 11 - 12 1753 104
XM_005257604.2 XP_005257661.2 11 - 12 1300 104
XM_017024967.1 XP_016880456.1 11 - 12 2492 104
GenBank mRNA AF113019.1 - 1 - 2 89 104
AK025917.1 - 3 - 4 317 104
AK300939.1 - 11 - 12 1537 104
AK303951.1 - 11 - 12 1339 104
AK307944.1 - 11 - 12 1306 104
BC018953.2 - 1 - 2 77 104
BC136322.1 - 12 - 13 1622 104
BC142963.1 - 11 - 12 1702 104

View Full Product Details