Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THOC7
Entrez Gene ID
80145
Gene Name
THO complex 7
Gene Aliases
NIF3L1BP1, fSAP24, hTREX30
Chromosome Location
Chr.3: 63833870 - 63864478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025075.3 NP_079351.2 1 - 2 158 68
GenBank mRNA AI391685.1 - 1 - 2 138 68
AK027098.1 - 1 - 2 68 68
AK225110.1 - 1 - 2 46 68
BC065012.1 - 1 - 2 28 68

View Full Product Details