The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013318.3 | NP_037450.2 | 26 - 27 | 6037 | 81 |
GenBank mRNA | AB011087.1 | - | 10 - 11 | 1306 | 81 |
AK026162.1 | - | 3 - 4 | 337 | 81 | |
BC012289.1 | - | 4 - 5 | 529 | 81 | |
BX647528.1 | - | 13 - 14 | 81 | ||
BX647842.1 | - | 12 - 13 | 1740 | 81 | |
CR749818.1 | - | 21 - 22 | 5112 | 81 | |
CR933661.1 | - | 14 - 15 | 3923 | 81 |