The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290474.1 | NP_001277403.1 | 2 - 3 | 844 | 92 |
NM_014807.4 | NP_055622.3 | 2 - 3 | 844 | 92 | |
XM_006718948.3 | XP_006719011.1 | 2 - 3 | 1133 | 92 | |
XM_011543076.2 | XP_011541378.1 | 2 - 3 | 1340 | 92 | |
XM_017018625.1 | XP_016874114.1 | 2 - 3 | 1133 | 92 | |
XM_017018626.1 | XP_016874115.1 | 2 - 3 | 1133 | 92 | |
XM_017018627.1 | XP_016874116.1 | 2 - 3 | 1133 | 92 | |
XM_017018628.1 | XP_016874117.1 | 2 - 3 | 269 | 92 | |
GenBank mRNA | AB006623.2 | - | 2 - 3 | 309 | 92 |
AB094094.1 | - | 2 - 3 | 812 | 92 | |
AK025223.1 | - | 2 - 3 | 812 | 92 | |
BC022219.1 | - | 2 - 3 | 710 | 92 |