The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290474.1 | NP_001277403.1 | 3 - 4 | 967 | 86 |
NM_014807.4 | NP_055622.3 | 3 - 4 | 967 | 86 | |
XM_011543076.2 | XP_011541378.1 | 3 - 4 | 1463 | 86 | |
XM_017018626.1 | XP_016874115.1 | 3 - 4 | 1256 | 86 | |
XM_017018627.1 | XP_016874116.1 | 3 - 4 | 1256 | 86 | |
XM_017018628.1 | XP_016874117.1 | 3 - 4 | 392 | 86 | |
GenBank mRNA | AB006623.2 | - | 3 - 4 | 432 | 86 |
AB094094.1 | - | 3 - 4 | 935 | 86 | |
AK025223.1 | - | 3 - 4 | 935 | 86 | |
BC022219.1 | - | 3 - 4 | 833 | 86 |