The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290474.1 | NP_001277403.1 | 4 - 5 | 1069 | 100 |
NM_014807.4 | NP_055622.3 | 4 - 5 | 1069 | 100 | |
XM_011543076.2 | XP_011541378.1 | 4 - 5 | 1565 | 100 | |
XM_017018626.1 | XP_016874115.1 | 4 - 5 | 1358 | 100 | |
XM_017018627.1 | XP_016874116.1 | 4 - 5 | 1358 | 100 | |
XM_017018628.1 | XP_016874117.1 | 4 - 5 | 494 | 100 | |
GenBank mRNA | AB006623.2 | - | 4 - 5 | 534 | 100 |
AB094094.1 | - | 4 - 5 | 1037 | 100 | |
AK025223.1 | - | 4 - 5 | 1037 | 100 | |
BC022219.1 | - | 4 - 5 | 935 | 100 | |
BX537572.1 | - | 1 - 2 | 370 | 100 |