The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290474.1 | NP_001277403.1 | 6 - 7 | 1307 | 61 |
NM_014807.4 | NP_055622.3 | 6 - 7 | 1307 | 61 | |
XM_006718948.3 | XP_006719011.1 | 5 - 6 | 1485 | 61 | |
XM_011543076.2 | XP_011541378.1 | 6 - 7 | 1803 | 61 | |
XM_017018625.1 | XP_016874114.1 | 5 - 6 | 1485 | 61 | |
XM_017018626.1 | XP_016874115.1 | 6 - 7 | 1596 | 61 | |
XM_017018627.1 | XP_016874116.1 | 6 - 7 | 1596 | 61 | |
XM_017018628.1 | XP_016874117.1 | 6 - 7 | 732 | 61 | |
GenBank mRNA | AB006623.2 | - | 6 - 7 | 772 | 61 |
AB094094.1 | - | 6 - 7 | 1275 | 61 | |
AK025223.1 | - | 6 - 7 | 1275 | 61 | |
BC022219.1 | - | 6 - 7 | 1173 | 61 | |
BX537572.1 | - | 3 - 4 | 608 | 61 |