The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290474.1 | NP_001277403.1 | 8 - 9 | 1470 | 68 |
NM_014807.4 | NP_055622.3 | 8 - 9 | 1470 | 68 | |
XM_006718948.3 | XP_006719011.1 | 7 - 8 | 1648 | 68 | |
XM_011543076.2 | XP_011541378.1 | 8 - 9 | 1966 | 68 | |
XM_017018625.1 | XP_016874114.1 | 7 - 8 | 1648 | 68 | |
XM_017018626.1 | XP_016874115.1 | 8 - 9 | 1759 | 68 | |
XM_017018627.1 | XP_016874116.1 | 8 - 9 | 1759 | 68 | |
XM_017018628.1 | XP_016874117.1 | 8 - 9 | 895 | 68 | |
GenBank mRNA | AB006623.2 | - | 8 - 9 | 935 | 68 |
AB094094.1 | - | 8 - 9 | 1438 | 68 | |
AK025223.1 | - | 8 - 9 | 1438 | 68 | |
BC010071.2 | - | 2 - 3 | 163 | 68 | |
BC021254.2 | - | 2 - 3 | 163 | 68 | |
BC022219.1 | - | 8 - 9 | 1336 | 68 | |
BC033083.1 | - | 2 - 3 | 163 | 68 | |
BC075832.1 | - | 2 - 3 | 163 | 68 | |
BX537572.1 | - | 4 - 5 | 1468 | 68 |