The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014807.4 | NP_055622.3 | 11 - 12 | 1882 | 80 |
XM_006718948.3 | XP_006719011.1 | 10 - 11 | 2060 | 80 | |
XM_011543076.2 | XP_011541378.1 | 11 - 12 | 2378 | 80 | |
XM_017018626.1 | XP_016874115.1 | 10 - 11 | 1970 | 80 | |
GenBank mRNA | AB094094.1 | - | 11 - 12 | 1850 | 80 |
AK025223.1 | - | 11 - 12 | 1850 | 80 | |
BC052246.1 | - | 3 - 4 | 352 | 80 |