The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006904.6 | NP_008835.5 | 80 - 81 | 11550 | 127 |
GenBank mRNA | BC037968.1 | - | 1 - 2 | 121 | 127 |
U47077.5 | - | 80 - 81 | 11550 | 127 |