The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128617.2 | NP_001122089.1 | 8 - 9 | 1710 | 73 |
NM_001286611.1 | NP_001273540.1 | 8 - 9 | 1710 | 73 | |
NM_001286612.1 | NP_001273541.1 | 8 - 9 | 1710 | 73 | |
NM_031922.4 | NP_114128.3 | 8 - 9 | 1710 | 73 | |
XM_005267177.3 | XP_005267234.1 | 8 - 9 | 1731 | 73 | |
XM_005267178.4 | XP_005267235.1 | 8 - 9 | 1731 | 73 | |
XM_005267179.3 | XP_005267236.1 | 8 - 9 | 1731 | 73 | |
XM_011536202.2 | XP_011534504.1 | 8 - 9 | 1097 | 73 | |
XM_017011387.1 | XP_016866876.1 | 8 - 9 | 1731 | 73 | |
XM_017011388.1 | XP_016866877.1 | 8 - 9 | 1117 | 73 | |
XM_017011389.1 | XP_016866878.1 | 8 - 9 | 1097 | 73 | |
GenBank mRNA | AB209143.1 | - | 5 - 6 | 1098 | 73 |
AF251052.1 | - | 8 - 9 | 1068 | 73 | |
AK312938.1 | - | 9 - 10 | 1406 | 73 | |
AL831900.1 | - | 1 - 2 | 158 | 73 | |
AL832307.1 | - | 7 - 8 | 956 | 73 | |
BC012764.2 | - | 8 - 9 | 1710 | 73 | |
BC021211.2 | - | 8 - 9 | 1330 | 73 |