The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303412.1 | NP_001290341.1 | 8 - 9 | 1244 | 74 |
XM_011526048.1 | XP_011524350.1 | 7 - 8 | 1166 | 74 | |
GenBank mRNA | BC000225.1 | - | 8 - 9 | 1180 | 74 |