The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004482.3 | NP_004473.2 | 5 - 6 | 1457 | 140 |
XM_005246449.1 | XP_005246506.1 | 5 - 6 | 1605 | 140 | |
XM_011510929.1 | XP_011509231.1 | 5 - 6 | 1340 | 140 | |
XM_017003770.1 | XP_016859259.1 | 5 - 6 | 3206 | 140 | |
GenBank mRNA | AK307391.1 | - | 6 - 7 | 1632 | 140 |
BC113565.1 | - | 4 - 5 | 1098 | 140 | |
BC113567.1 | - | 4 - 5 | 1098 | 140 | |
BX647473.1 | - | 2 - 3 | 297 | 140 | |
X92689.1 | - | 4 - 5 | 1070 | 140 |