Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIN2C
Entrez Gene ID
2905
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
Gene Aliases
GluN2C, NMDAR2C, NR2C
Chromosome Location
Chr.17: 74842021 - 74860843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000835.4 NP_000826.2 10 - 11 2320 103
NM_001278553.1 NP_001265482.1 10 - 11 2574 103
XM_006721845.3 XP_006721908.1 9 - 10 3586 103
XM_006721846.3 XP_006721909.1 9 - 10 3584 103
XM_011524687.2 XP_011522989.1 10 - 11 3651 103
GenBank mRNA AB208799.1 - 6 - 7 5909 103
BC041128.1 - 10 - 11 2266 103
BC140801.1 - 10 - 11 2198 103
L76224.1 - 9 - 10 2164 103
U77782.1 - 10 - 11 2352 103

View Full Product Details