Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIN2C
Entrez Gene ID
2905
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
Gene Aliases
GluN2C, NMDAR2C, NR2C
Chromosome Location
Chr.17: 74842021 - 74860843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000835.4 NP_000826.2 11 - 12 2510 85
NM_001278553.1 NP_001265482.1 11 - 12 2764 85
NR_103735.1 11 - 12 2506 85
XM_006721845.3 XP_006721908.1 10 - 11 3776 85
XM_006721846.3 XP_006721909.1 10 - 11 3774 85
XM_011524686.2 XP_011522988.1 11 - 12 3865 85
XM_011524687.2 XP_011522989.1 11 - 12 3841 85
XM_011524688.2 XP_011522990.1 10 - 11 3801 85
XM_011524689.2 XP_011522991.1 12 - 13 2588 85
XM_011524692.2 XP_011522994.1 11 - 12 3787 85
XM_017024544.1 XP_016880033.1 11 - 12 3866 85
GenBank mRNA AB208799.1 - 7 - 8 6099 85
BC041128.1 - 11 - 12 2456 85
BC140801.1 - 11 - 12 2388 85
L76224.1 - 10 - 11 2354 85
U77782.1 - 11 - 12 2542 85

View Full Product Details