Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCGF1
Entrez Gene ID
84759
Gene Name
polycomb group ring finger 1
Gene Aliases
2010002K04Rik, NSPC1, RNF3A-2, RNF68
Chromosome Location
Chr.2: 74505043 - 74508580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032673.2 NP_116062.2 7 - 8 680 77
GenBank mRNA AF087884.1 - 6 - 7 786 77
AK125742.1 - 7 - 8 1566 77
AL049296.1 - 1 - 2 720 77
BC004952.1 - 7 - 8 645 77

View Full Product Details