The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013261.3 | NP_037393.1 | 3 - 4 | 546 | 96 |
XM_005248131.4 | XP_005248188.1 | 3 - 4 | 1289 | 96 | |
XM_005248132.1 | XP_005248189.1 | 3 - 4 | 468 | 96 | |
XM_005248134.4 | XP_005248191.1 | 3 - 4 | 535 | 96 | |
XM_011513765.2 | XP_011512067.1 | 4 - 5 | 1038 | 96 | |
XM_011513766.1 | XP_011512068.1 | 4 - 5 | 620 | 96 | |
XM_011513767.2 | XP_011512069.1 | 4 - 5 | 2758 | 96 | |
XM_011513768.1 | XP_011512070.1 | 3 - 4 | 519 | 96 | |
XM_011513769.2 | XP_011512071.1 | 3 - 4 | 535 | 96 | |
XM_011513770.2 | XP_011512072.1 | 2 - 3 | 756 | 96 | |
XM_011513771.1 | XP_011512073.1 | 2 - 3 | 365 | 96 | |
XM_017007664.1 | XP_016863153.1 | 5 - 6 | 995 | 96 | |
GenBank mRNA | AB061325.1 | - | 3 - 4 | 501 | 96 |
AF106698.1 | - | 3 - 4 | 546 | 96 | |
AF159714.1 | - | 3 - 4 | 455 | 96 | |
AF186379.1 | - | 3 - 4 | 465 | 96 | |
AK296591.1 | - | 3 - 4 | 504 | 96 | |
AK301883.1 | - | 3 - 4 | 498 | 96 | |
AK309261.1 | - | 3 - 4 | 504 | 96 | |
HQ695733.1 | - | 2 - 3 | 756 | 96 | |
JQ772116.1 | - | 4 - 5 | 834 | 96 | |
JQ772117.1 | - | 4 - 5 | 937 | 96 | |
JQ772118.1 | - | 5 - 6 | 1002 | 96 | |
JQ772119.1 | - | 4 - 5 | 834 | 96 |