The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319116.1 | NP_001306045.1 | 4 - 5 | 459 | 102 |
NM_001319117.1 | NP_001306046.1 | 4 - 5 | 459 | 102 | |
NM_001319118.1 | NP_001306047.1 | 4 - 5 | 387 | 102 | |
NM_001319119.1 | NP_001306048.1 | 4 - 5 | 667 | 102 | |
NM_001319120.1 | NP_001306049.1 | 4 - 5 | 484 | 102 | |
NM_001319121.1 | NP_001306050.1 | 5 - 6 | 360 | 102 | |
NM_001319122.1 | NP_001306051.1 | 4 - 5 | 452 | 102 | |
NM_012100.3 | NP_036232.2 | 4 - 5 | 484 | 102 | |
NR_134970.1 | 4 - 5 | 484 | 102 | ||
GenBank mRNA | AF005050.2 | - | 4 - 5 | 459 | 102 |
AK001777.1 | - | 4 - 5 | 484 | 102 | |
AK001977.1 | - | 4 - 5 | 456 | 102 | |
AK021871.1 | - | 4 - 5 | 336 | 102 | |
AK023005.1 | - | 4 - 5 | 398 | 102 | |
AK301919.1 | - | 4 - 5 | 444 | 102 | |
AK302809.1 | - | 4 - 5 | 451 | 102 | |
AL519865.3 | - | 4 - 5 | 430 | 102 | |
BC000653.2 | - | 4 - 5 | 366 | 102 | |
BI256589.1 | - | 4 - 5 | 349 | 102 | |
BI821651.1 | - | 4 - 5 | 372 | 102 | |
BM564135.1 | - | 4 - 5 | 447 | 102 | |
BX403948.2 | - | 4 - 5 | 516 | 102 | |
CN356162.1 | - | 4 - 5 | 378 | 102 | |
DA203030.1 | - | 4 - 5 | 484 | 102 | |
DA398660.1 | - | 4 - 5 | 387 | 102 |