The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286426.1 | NP_001273355.1 | 10 - 11 | 1328 | 101 |
NM_001286428.1 | NP_001273357.1 | 9 - 10 | 1218 | 101 | |
NM_003800.4 | NP_003791.3 | 10 - 11 | 1328 | 101 | |
XM_017011401.1 | XP_016866890.1 | 10 - 11 | 1415 | 101 | |
XM_017011402.1 | XP_016866891.1 | 10 - 11 | 1415 | 101 | |
GenBank mRNA | AB009022.1 | - | 10 - 11 | 1108 | 101 |
AB009023.1 | - | 10 - 11 | 1108 | 101 | |
AB009024.1 | - | 10 - 11 | 1108 | 101 | |
AB012142.1 | - | 10 - 11 | 1263 | 101 | |
AB012143.1 | - | 10 - 11 | 1263 | 101 | |
AF025654.1 | - | 10 - 11 | 1396 | 101 | |
AK295719.1 | - | 11 - 12 | 1206 | 101 | |
AK299777.1 | - | 9 - 10 | 1208 | 101 | |
AK312407.1 | - | 10 - 11 | 1299 | 101 | |
BC019954.1 | - | 10 - 11 | 1295 | 101 |