Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNGTT
Entrez Gene ID
8732
Gene Name
RNA guanylyltransferase and 5'-phosphatase
Gene Aliases
CAP1A, HCE, HCE1, hCAP
Chromosome Location
Chr.6: 88609897 - 88963716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286426.1 NP_001273355.1 3 - 4 501 145
NM_001286428.1 NP_001273357.1 2 - 3 391 145
NM_003800.4 NP_003791.3 3 - 4 501 145
XM_017011401.1 XP_016866890.1 3 - 4 588 145
XM_017011402.1 XP_016866891.1 3 - 4 588 145
GenBank mRNA AB009022.1 - 3 - 4 281 145
AB009023.1 - 3 - 4 281 145
AB009024.1 - 3 - 4 281 145
AB012142.1 - 3 - 4 436 145
AB012143.1 - 3 - 4 436 145
AF025654.1 - 3 - 4 569 145
AK295719.1 - 4 - 5 379 145
AK299777.1 - 2 - 3 381 145
AK312407.1 - 3 - 4 472 145
BC019954.1 - 3 - 4 468 145
BP256840.1 - 3 - 4 501 145

View Full Product Details