Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM23
Entrez Gene ID
55147
Gene Name
RNA binding motif protein 23
Gene Aliases
CAPERbeta, PP239, RNPC4
Chromosome Location
Chr.14: 22900645 - 22919187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077352.1 NP_001070820.1 3 - 4 378 80
NM_018107.4 NP_060577.3 3 - 4 378 80
XM_011536892.2 XP_011535194.1 4 - 5 460 80
XM_011536893.2 XP_011535195.1 4 - 5 461 80
XM_011536895.2 XP_011535197.1 4 - 5 485 80
XM_011536896.2 XP_011535198.1 4 - 5 461 80
XM_011536897.2 XP_011535199.1 3 - 4 793 80
XM_017021398.1 XP_016876887.1 4 - 5 463 80
XM_017021399.1 XP_016876888.1 4 - 5 464 80
XM_017021400.1 XP_016876889.1 3 - 4 328 80
XM_017021403.1 XP_016876892.1 3 - 4 352 80
XM_017021404.1 XP_016876893.1 3 - 4 302 80
XM_017021408.1 XP_016876897.1 3 - 4 340 80
XM_017021410.1 XP_016876899.1 3 - 4 362 80
XM_017021411.1 XP_016876900.1 3 - 4 793 80
XM_017021412.1 XP_016876901.1 4 - 5 461 80
GenBank mRNA AF087905.1 - 3 - 4 329 80
AK001344.1 - 3 - 4 328 80
AK295448.1 - 3 - 4 340 80
AK308055.1 - 3 - 4 341 80
AK315620.1 - 3 - 4 340 80
BC002566.2 - 3 - 4 356 80
BC024208.2 - 3 - 4 315 80
CK000110.1 - 3 - 4 352 80
CR457257.1 - 2 - 3 179 80
DC363343.1 - 3 - 4 373 80
KU178687.1 - 2 - 3 179 80
KU178689.1 - 2 - 3 179 80

View Full Product Details