Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EPHA10
Entrez Gene ID
284656
Gene Name
EPH receptor A10
Gene Aliases
-
Chromosome Location
Chr.1: 37713888 - 37765152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099439.1 NP_001092909.1 9 - 10 1925 128
XM_017001080.1 XP_016856569.1 5 - 6 667 128
XM_017001081.1 XP_016856570.1 5 - 6 667 128
GenBank mRNA AK131215.1 - 5 - 6 639 128
AK309089.1 - 5 - 6 577 128
AY726560.1 - 3 - 4 260 128
BC041360.1 - 5 - 6 513 128
BC112933.1 - 3 - 4 339 128

View Full Product Details