Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRGAP2
Entrez Gene ID
23380
Gene Name
SLIT-ROBO Rho GTPase activating protein 2
Gene Aliases
ARHGAP34, FNBP2, SRGAP2A, SRGAP3
Chromosome Location
Chr.1: 206203464 - 206464443 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170637.3 NP_001164108.1 11 - 12 2009 63
NM_001300952.1 NP_001287881.1 11 - 12 2009 63
NM_015326.4 NP_056141.2 11 - 12 2012 63
XM_005277510.2 XP_005277567.1 12 - 13 2199 63
XM_005277511.2 XP_005277568.1 12 - 13 2196 63
XM_005277514.1 XP_005277571.1 12 - 13 2199 63
XM_005277515.1 XP_005277572.1 12 - 13 2196 63
XM_011509354.1 XP_011507656.1 12 - 13 2199 63
XM_011509355.1 XP_011507657.1 12 - 13 2196 63
XM_011509356.2 XP_011507658.1 12 - 13 2199 63
XM_011509359.1 XP_011507661.1 12 - 13 2199 63
XM_017000838.1 XP_016856327.1 10 - 11 1843 63
XM_017000839.1 XP_016856328.1 10 - 11 1873 63
XM_017000840.1 XP_016856329.1 12 - 13 1488 63
XM_017000841.1 XP_016856330.1 12 - 13 2196 63
GenBank mRNA AB007925.1 - 11 - 12 1542 63
AK293335.1 - 8 - 9 1048 63
AK295845.1 - 11 - 12 1765 63
BC041635.1 - 3 - 4 313 63
BC132872.1 - 11 - 12 1540 63
BC132874.1 - 11 - 12 1540 63
BC144343.1 - 11 - 12 1537 63

View Full Product Details