Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PTPN4
- Entrez Gene ID
-
5775
- Gene Name
- protein tyrosine phosphatase, non-receptor type 4
- Gene Aliases
-
MEG, PTPMEG, PTPMEG1
- Chromosome Location
-
Chr.2: 119759631 - 119984899 on Build GRCh38
View Full Product Details