The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004140.3 | NP_004131.3 | 20 - 21 | 3051 | 121 |
XM_011523853.2 | XP_011522155.1 | 20 - 21 | 3080 | 121 | |
XM_011523855.2 | XP_011522157.1 | 20 - 21 | 3074 | 121 | |
XM_011523856.1 | XP_011522158.1 | 20 - 21 | 3068 | 121 | |
GenBank mRNA | BC151838.1 | - | 20 - 21 | 3027 | 121 |
BQ923585.1 | - | 3 - 4 | 121 | ||
X86371.1 | - | 20 - 21 | 2984 | 121 |