The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004140.3 | NP_004131.3 | 6 - 7 | 775 | 88 |
XM_011523849.2 | XP_011522151.1 | 6 - 7 | 804 | 88 | |
XM_011523851.2 | XP_011522153.1 | 6 - 7 | 804 | 88 | |
XM_011523852.2 | XP_011522154.1 | 6 - 7 | 804 | 88 | |
XM_011523853.2 | XP_011522155.1 | 6 - 7 | 804 | 88 | |
XM_011523854.2 | XP_011522156.1 | 6 - 7 | 804 | 88 | |
XM_011523855.2 | XP_011522157.1 | 6 - 7 | 804 | 88 | |
XM_011523856.1 | XP_011522158.1 | 6 - 7 | 804 | 88 | |
GenBank mRNA | BC028037.1 | - | 6 - 7 | 810 | 88 |
BC028214.1 | - | 6 - 7 | 749 | 88 | |
BC151838.1 | - | 6 - 7 | 751 | 88 | |
X86371.1 | - | 6 - 7 | 723 | 88 |