The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006310.3 | NP_006301.3 | 21 - 22 | 2788 | 81 |
XM_011525496.1 | XP_011523798.1 | 22 - 23 | 2590 | 81 | |
GenBank mRNA | AJ132583.1 | - | 21 - 22 | 2518 | 81 |
AK293450.1 | - | 15 - 16 | 1720 | 81 | |
AK293995.1 | - | 14 - 15 | 1702 | 81 | |
AK296887.1 | - | 20 - 21 | 2431 | 81 | |
AK297107.1 | - | 20 - 21 | 2292 | 81 | |
AK303037.1 | - | 22 - 23 | 2583 | 81 | |
BC065294.1 | - | 21 - 22 | 2477 | 81 |