The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173561.2 | NP_775832.2 | 3 - 4 | 783 | 104 |
GenBank mRNA | AY510109.1 | - | 2 - 3 | 684 | 104 |
BC035284.1 | - | 3 - 4 | 773 | 104 | |
JF681947.1 | - | 3 - 4 | 801 | 104 |