The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173561.2 | NP_775832.2 | 6 - 7 | 1214 | 105 |
GenBank mRNA | AY510109.1 | - | 5 - 6 | 1115 | 105 |
BC035284.1 | - | 6 - 7 | 1204 | 105 | |
BM312066.1 | - | 3 - 4 | 256 | 105 | |
JF681947.1 | - | 6 - 7 | 1232 | 105 |