Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SENP3
- Entrez Gene ID
-
26168
- Gene Name
- SUMO1/sentrin/SMT3 specific peptidase 3
- Gene Aliases
-
SMT3IP1, SSP3, Ulp1
- Chromosome Location
-
Chr.17: 7561992 - 7571969 on Build GRCh38
- Gene Symbol
- SENP3-EIF4A1
- Entrez Gene ID
-
100533955
- Gene Name
- SENP3-EIF4A1 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 7561992 - 7579006 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037926.1 |
|
6 - 7 |
1544 |
86 |
View Full Product Details