The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075098.3 | NP_001068566.1 | 4 - 5 | 720 | 76 |
NM_005943.5 | NP_005934.2 | 3 - 4 | 724 | 76 | |
NR_033233.1 | 3 - 4 | 593 | 76 | ||
XM_011514632.2 | XP_011512934.1 | 4 - 5 | 631 | 76 | |
XM_011514633.2 | XP_011512935.1 | 4 - 5 | 631 | 76 | |
XM_011514634.2 | XP_011512936.1 | 3 - 4 | 528 | 76 | |
XM_017010888.1 | XP_016866377.1 | 3 - 4 | 538 | 76 | |
GenBank mRNA | AF034374.1 | - | 4 - 5 | 603 | 76 |
AJ224328.1 | - | 3 - 4 | 721 | 76 | |
AK056740.1 | - | 3 - 4 | 539 | 76 | |
AK091306.1 | - | 3 - 4 | 452 | 76 | |
AK300300.1 | - | 4 - 5 | 593 | 76 | |
BC036839.1 | - | 4 - 5 | 593 | 76 |