Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOCS1
Entrez Gene ID
4337
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
MIG11, MOCOD, MOCODA
Chromosome Location
Chr.6: 39904258 - 39934551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033233.1 1 - 2 254 118
XM_011514634.2 XP_011512936.1 1 - 2 189 118
XM_017010888.1 XP_016866377.1 1 - 2 199 118
GenBank mRNA AK056740.1 - 1 - 2 200 118

View Full Product Details