The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079866.1 | NP_001073335.1 | 6 - 7 | 1023 | 69 |
NM_001257342.1 | NP_001244271.1 | 7 - 8 | 1153 | 69 | |
NM_001257343.1 | NP_001244272.1 | 7 - 8 | 1205 | 69 | |
NM_001257344.1 | NP_001244273.1 | 6 - 7 | 1080 | 69 | |
NM_001318836.1 | NP_001305765.1 | 5 - 6 | 654 | 69 | |
NM_001320717.1 | NP_001307646.1 | 7 - 8 | 1195 | 69 | |
NM_004328.4 | NP_004319.1 | 7 - 8 | 1232 | 69 | |
XM_005246748.2 | XP_005246805.1 | 6 - 7 | 1507 | 69 | |
XM_005246749.3 | XP_005246806.1 | 4 - 5 | 506 | 69 | |
XM_006712678.1 | XP_006712741.1 | 6 - 7 | 1638 | 69 | |
XM_017004631.1 | XP_016860120.1 | 7 - 8 | 1394 | 69 | |
XM_017004632.1 | XP_016860121.1 | 7 - 8 | 1119 | 69 | |
XM_017004633.1 | XP_016860122.1 | 6 - 7 | 1897 | 69 | |
XM_017004634.1 | XP_016860123.1 | 5 - 6 | 925 | 69 | |
GenBank mRNA | AF026849.1 | - | 6 - 7 | 999 | 69 |
AF038195.1 | - | 6 - 7 | 961 | 69 | |
AK096210.1 | - | 6 - 7 | 1586 | 69 | |
AK223518.1 | - | 6 - 7 | 1109 | 69 | |
AK289363.1 | - | 7 - 8 | 1095 | 69 | |
BC000416.2 | - | 6 - 7 | 1153 | 69 | |
BC007500.2 | - | 6 - 7 | 1153 | 69 | |
BX571752.1 | - | 7 - 8 | 1152 | 69 | |
KU177941.1 | - | 5 - 6 | 886 | 69 | |
KU177942.1 | - | 4 - 5 | 741 | 69 |