The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126131.1 | NP_001119603.1 | 10 - 11 | 2219 | 148 |
NM_002693.2 | NP_002684.1 | 10 - 11 | 2231 | 148 | |
GenBank mRNA | AK291281.1 | - | 10 - 11 | 2250 | 148 |
BC042571.1 | - | 10 - 11 | 2216 | 148 | |
BC050559.1 | - | 10 - 11 | 2200 | 148 | |
D84103.1 | - | 9 - 10 | 2055 | 148 | |
U60325.1 | - | 9 - 10 | 2108 | 148 | |
X98093.1 | - | 10 - 11 | 2231 | 148 |