The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126131.1 | NP_001119603.1 | 11 - 12 | 2338 | 76 |
NM_002693.2 | NP_002684.1 | 11 - 12 | 2350 | 76 | |
GenBank mRNA | AK291281.1 | - | 11 - 12 | 2369 | 76 |
BC042571.1 | - | 11 - 12 | 2335 | 76 | |
BC050559.1 | - | 11 - 12 | 2319 | 76 | |
D84103.1 | - | 10 - 11 | 2174 | 76 | |
U60325.1 | - | 10 - 11 | 2227 | 76 | |
X98093.1 | - | 11 - 12 | 2350 | 76 |