The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126131.1 | NP_001119603.1 | 6 - 7 | 1517 | 55 |
NM_002693.2 | NP_002684.1 | 6 - 7 | 1529 | 55 | |
GenBank mRNA | AK291281.1 | - | 6 - 7 | 1548 | 55 |
BC042571.1 | - | 6 - 7 | 1514 | 55 | |
BC050559.1 | - | 6 - 7 | 1498 | 55 | |
D84103.1 | - | 5 - 6 | 1353 | 55 | |
U60325.1 | - | 5 - 6 | 1406 | 55 | |
X98093.1 | - | 6 - 7 | 1529 | 55 |